missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Bitartrate Dietary Supplement, USP, 99-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Choline Bitartrate, USP Dietary Supplement is an essential nutrient and typically grouped with B-complex vitamins. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.1% |
| Packaging | Poly Pail |
| Quantity | 2.5 kg |
| Optical Rotation | +17.5 to +18.5° |
| Percent Purity | 99-100.5% |
| Grade | USP Dietary Supplement |