missing translation for 'onlineSavingsMsg'
Learn More

Choline Bitartrate Dietary Supplement, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. 18601670
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601670 500 g Poly Bottle
18601669 125 g Poly Bottle
18601671 2.5 kg Poly Pail
18601672 12 kg Poly Pail
18601673 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601670 Supplier Spectrum Chemical Mfg Cor Supplier No. C2052500GM
Only null left

C2052, 87-67-2, C9H19NO7

Spectrum™ Chemical Choline Bitartrate, USP Dietary Supplement is an essential nutrient and typically grouped with B-complex vitamins. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-67-2
Molecular Formula C9H19NO7
Molecular Weight (g/mol) 253.25
InChI Key QWJSAWXRUVVRLH-LREBCSMRSA-M
IUPAC Name (2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
SMILES C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 500 g
Optical Rotation +17.5 to +18.5°
Percent Purity 99-100.5%
Grade USP Dietary Supplement