missing translation for 'onlineSavingsMsg'
Learn More

Choline bitartrate, 98+%

CAS: 87-67-2 | C9H19NO7 | 253.251 g/mol

Supplier:  Thermo Scientific Chemicals J64657A1

 View more versions of this product

Catalog No. AAJ64657A1


Only null left
Explore available promotions

Description

Description

Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Choline bitartrate
151°C to 153°C
C9H19NO7
1 kg
choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
QWJSAWXRUVVRLH-UHFFFAOYSA-M
2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
10198924
≥98%
87-67-2
White
MFCD00036332
Hygroscopic
Soluble in alcohol and slightly soluble in water.
C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O
253.251
253.25
Powder

Chemical Identifiers

87-67-2
253.251
QWJSAWXRUVVRLH-UHFFFAOYSA-M
10198924
C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O
C9H19NO7
MFCD00036332
choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 201-763-4

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only