missing translation for 'onlineSavingsMsg'
Learn More

Choline bitartrate, 98+%

Catalog No. AAJ64657A4
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64657A4 2.5 kg
AAJ6465730 250 g
AAJ64657A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ64657A4 Supplier Thermo Scientific Chemicals Supplier No. J64657A4

missing translation for 'validMainframeMsg'

CAS: 87-67-2 | C9H19NO7 | 253.251 g/mol

Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 87-67-2
Molecular Formula C9H19NO7
Molecular Weight (g/mol) 253.251
MDL Number MFCD00036332
InChI Key QWJSAWXRUVVRLH-UHFFFAOYSA-M
Synonym choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
PubChem CID 10198924
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
SMILES C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O

Specifications

Melting Point 151°C to 153°C
Color White
Quantity 2.5 kg
Sensitivity Hygroscopic
Solubility Information Soluble in alcohol and slightly soluble in water.
Formula Weight 253.25
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Choline bitartrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-763-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only