missing translation for 'onlineSavingsMsg'
Learn More

Choline bitartrate, 97%

Catalog No. AC450221000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC450221000 100 g Glass bottle
AC450225000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450221000 Supplier Thermo Scientific Chemicals Supplier No. 450221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 87-67-2 | C5H14NO·C4H5O6 | 253.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-67-2
Molecular Formula C5H14NO·C4H5O6
Molecular Weight (g/mol) 253.25
InChI Key QWJSAWXRUVVRLH-UHFFFAOYSA-M
Synonym choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
PubChem CID 10198924
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
SMILES C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O

Specifications

Melting Point 151.0°C to 153.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Merck Index 15,2211
Solubility Information Solubility in water: soluble.
Formula Weight 253.25
Percent Purity 97%
Chemical Name or Material Choline bitartrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 201-763-4

RUO – Research Use Only