missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl nonanoate, Thermo Scientific Chemicals

Catalog No. AAL0285722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0285722 100 g
AAL0285714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0285722 Supplier Thermo Scientific Chemicals Supplier No. L0285722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cholesteryl nonanoate is used in hair colors, cosmetic products like lip products, lotions. It is also used in the manufature of some pleochroic dyes. It can be also used as a component of the liquid crystals used for liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl nonanoate is used in hair colors, cosmetic products like lip products, lotions. It is also used in the manufature of some pleochroic dyes. It can be also used as a component of the liquid crystals used for liquid crystal displays.

Solubility
Soluble in Chloroform, Tetrahydrofuran(THF). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.

Chemical Identifiers

CAS 1182-66-7
Molecular Formula C36H62O2
Molecular Weight (g/mol) 526.89
MDL Number MFCD00003643
InChI Key WCLNGBQPTVENHV-MKQVXYPISA-N
Synonym cholesteryl pelargonate, cholesteryl nonanoate, cholesterol pelargonate, unii-4313o7p4xw, 5-cholesten-3beta-ol 3-nonanoate, cholesterol, nonanoate, 3beta-hydroxy-5-cholestene 3-nonanoate, nonanoic acid cholesterol ester, pelargonic acid cholesterol ester, cholest-5-ene-3-beta-yl nonanoate
PubChem CID 2723614
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] nonanoate
SMILES CCCCCCCCC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CCC4C(C)CCCC(C)C)C)C

Specifications

Melting Point 75°C to 79°C
Odor Odorless
Quantity 100 g
Beilstein 3179820
Solubility Information Soluble in Chloroform,Tetrahydrofuran(THF). Insoluble in water.
Optical Rotation −30° (c=5 in Chloroform)
Formula Weight 526.89
Chemical Name or Material Cholesteryl nonanoate
EINECSNumber 214-658-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only