missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cholecystokinin, CCK Octapeptide (26-33)
Description
Cholecystokinin, CCK Octapeptide (26-33) is used in the biological studies in the preparation of opioid receptors agonistic and CCK receptor antagonistic activity. It plays a role in satiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Purification Method | HPLC |
| Common Name | CCK 8 |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 1143.27 |
| Product Type | Cholecystokinin, CCK Octapeptide (26-33) |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Sequence | Asp-Tyr(SO3H)-Met-Gly-Trp-Met-Asp-Phe-NH2 |
RUO – Research Use Only