missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cholecystokinin, CCK (27-33)
For cellular and molecular biology applications
$127.10 - $127.10
Specifications
| Purification Method | HPLC |
|---|---|
| Common Name | CCK Heptapeptide |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 948.12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ66940MCR
|
Thermo Scientific Chemicals
J66940MCR |
1 mg |
Each for $127.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| HPLC | |
| -30°C to -10°C | |
| 948.12 | |
| RUO - research use only |
| CCK Heptapeptide | |
| Solid | |
| Cholecystokinin, CCK (27-33) | |
| Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2 |
RUO – Research Use Only