missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CHO DG44 Cells (cGMP banked) and Media Kit

Catalog No. A1100001
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1100001 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1100001 Supplier Gibco™ Supplier No. A1100001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHO DG44 Cells (cGMP banked) and Media Kit

The CHO DG44 Cells (cGMP banked) and Media Kit is optimized for the growth of dihydrofolate reductase deficient (DHFR-) Chinese Hamster Ovary (CHO) cells and expression of recombinant proteins in suspension culture. The kit provides:

Gibco™ CD DG44 Medium—a chemically defined, protein-free medium specifically designed to enable optimal growth and expression of DG44 cells in suspension culture

CHO DG44 cells (cGMP banked) —Parental CHO DG44 cells have been produced, banked and tested to meet cGMP quality standards. They are pre-adapted to CD DG44 Medium and selected for superior cell growth and transfection efficiencies.

In addition, the CHO DG44 Cells (cGMP banked) and Media Kit includes L-glutamine and Pluronic™ F-68 for optimal growth and protein expression of DG44 cells in a protein-free, chemically defined environment.

Specifications

Classification Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Content And Storage CHO DG44 Cells (cGMP banked) and Media Kit (SKU A1100001) contains:

• CD DG44 Medium 1000 mL- 12610-010, store in the dark at 2-8°C
• CHO DG44 Cells (cGMP banked) 1 vial (> 1x10^7 cells) - A1097101 (see limited label license), store in liquid nitrogen
• PLURONIC F-68, 10% (100X) 100 mL - 24040-032, store at room temperature
• L-glutamine-200mM (100X) 100 mL - 25030-081 or 25030-024 (CE marked version for Europe only), store at -20°C
Cell Line CHO DG44
Culture Type Suspension Cell Culture
Species Hamster
Media Recommendation CD DG44 Medium
No. of Cells 1 x 107
Product Line Gibco
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.