missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline hydrochloride

Catalog No. p-7041746
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6009514 25 g
AAJ6009522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6009514 Supplier Thermo Scientific Chemicals Supplier No. J6009514

Bacteriostatic antibiotic active against Gram-positive and Gram-negative bacteria | CAS: 64-72-2 | C22H24Cl2N2O8 | 515.34 g/mol

Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

Solubility
Soluble in water

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22H24Cl2N2O8
Molecular Weight (g/mol) 515.34
MDL Number MFCD00082440
InChI Key QYAPHLRPFNSDNH-CIVPRPTRSA-N
Synonym chlortetracycline hydrochloride, 4-epi-chlortetracycline hydrochloride, 2-amino hydroxy methylidene-7-chloro-4-dimethylamino-6,10,11,12a-tetrahydroxy-6-methyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride, 2-azanyl oxidanyl methylidene-7-chloranyl-4-dimethylamino-6-methyl-6,10,11,12a-tetrakis oxidanyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride
PubChem CID 66577600
IUPAC Name (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Melting Point 210°C to 215°C (decomposition)
Color Yellow to Brown
pH 2.3 to 3.3
Quantity 25 g
Beilstein 3858364
Sensitivity Light sensitive
Merck Index 14,2192
Solubility Information Soluble in water
Formula Weight 515.34
Physical Form Powder
Chemical Name or Material Chlortetracycline hydrochloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-591-7
RTECSNumber QI7800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only