missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline Hydrochloride, MP Biomedicals

Catalog No. ICN19032725
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19032725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19032725 Supplier MP Biomedicals Inc Supplier No. 19032725

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Antibacterial

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22H24Cl2N2O8
Molecular Weight (g/mol) 515.34
InChI Key QYAPHLRPFNSDNH-CIVPRPTRSA-N
PubChem CID 66577600
IUPAC Name (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Type Chlortetracycline Hydrochloride
Quantity 25 g
Formula Weight 515.3
Chemical Name or Material Chlortetracycline Hydrochloride