missing translation for 'onlineSavingsMsg'
Learn More

Chlorpromazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7125485
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C24815G 5 g
C248125G 25 g
2 options

Catalog No. C24815G

Supplier: TCI America C24815G

Only null left

Chemical Identifiers

CAS 69-09-0
Molecular Formula C17H20Cl2N2S
Molecular Weight (g/mol) 355.321
MDL Number MFCD00012654
InChI Key FBSMERQALIEGJT-UHFFFAOYSA-N
Synonym chlorpromazine hydrochloride, sonazine, chlorpromazine hcl, chloractil, klorpromex, promacid, hebanil, propaphenin, hibernal, hybernal
PubChem CID 6240
ChEBI CHEBI:3649
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2SC3=C1C=C(C=C3)Cl.Cl

Specifications

Melting Point 197°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 355.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorpromazine Hydrochloride
RTECSNumber SO1750000
TSCA Yes