missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine maleate, 99%

Catalog No. AAB2523806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2523806 5 g
AAB2523814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2523806 Supplier Thermo Scientific Chemicals Supplier No. B2523806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 113-92-8 | C20H23ClN2O4 | 390.86 g/mol

An antihistaminic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 113-92-8
Molecular Formula C20H23ClN2O4
Molecular Weight (g/mol) 390.86
MDL Number MFCD00069225
InChI Key DBAKFASWICGISY-BTJKTKAUNA-N
Synonym chlorpheniramine maleate, antagonate, allerclor, allergin, chlormene, alunex, carbinoxamide maleate, allergisan, -chlorpheniramine maleate salt, dl-2-p-chloro-a-2-dimethylamino ethylbenzyl pyridine bimaleate
PubChem CID 5281068
ChEBI CHEBI:3645
IUPAC Name [3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
SMILES OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1

Specifications

Melting Point 130°C to 135°C
Quantity 5 g
Formula Weight 390.86
Percent Purity 99%
Chemical Name or Material Chlorpheniramine maleate
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-037-5
RTECSNumber US6503000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only