missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotripyrrolidinophosphonium Hexafluorophosphate 98.0+%, TCI America™
$115.51 - $115.51
Chemical Identifiers
| CAS | 133894-48-1 |
|---|---|
| Molecular Formula | C12H24ClF6N3P2 |
| Molecular Weight (g/mol) | 421.733 |
| MDL Number | MFCD00210035 |
| InChI Key | BSCYRXJVGSZNKX-UHFFFAOYSA-N |
| Synonym | pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat |
| PubChem CID | 16211753 |
| IUPAC Name | chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate |
| SMILES | C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F |
Chemical Identifiers
| 133894-48-1 | |
| 421.733 | |
| BSCYRXJVGSZNKX-UHFFFAOYSA-N | |
| 16211753 | |
| C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F |
| C12H24ClF6N3P2 | |
| MFCD00210035 | |
| pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat | |
| chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate |
Specifications
| 133894-48-1 | |
| White-Yellow | |
| MFCD00210035 | |
| 1759 | |
| BSCYRXJVGSZNKX-UHFFFAOYSA-N | |
| chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate | |
| 16211753 | |
| ≥98.0% (HPLC,T) | |
| Chlorotripyrrolidinophosphonium Hexafluorophosphate |
| 138°C | |
| C12H24ClF6N3P2 | |
| 5 g | |
| pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat | |
| C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F | |
| 421.733 | |
| 421.73 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator