missing translation for 'onlineSavingsMsg'
Learn More

Chlorotripyrrolidinophosphonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. C25515G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25515G Supplier TCI America Supplier No. C25515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133894-48-1
Molecular Formula C12H24ClF6N3P2
Molecular Weight (g/mol) 421.733
MDL Number MFCD00210035
InChI Key BSCYRXJVGSZNKX-UHFFFAOYSA-N
Synonym pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat
PubChem CID 16211753
IUPAC Name chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 421.73
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorotripyrrolidinophosphonium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator