missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriphenylsilane, 96%

Catalog No. AAA1367806
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1367806 5 g
AAA1367814 25 g
AAA1367818 50 g
AAA1367822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1367806 Supplier Thermo Scientific Chemicals Supplier No. A1367806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 76-86-8 | C18H15ClSi | 294.85 g/mol

Chlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store under dry inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.

Chemical Identifiers

CAS 76-86-8
Molecular Formula C18H15ClSi
Molecular Weight (g/mol) 294.85
MDL Number MFCD00000496
InChI Key MNKYQPOFRKPUAE-UHFFFAOYSA-N
Synonym triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl
PubChem CID 6458
IUPAC Name chloro(triphenyl)silane
SMILES Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 188°C to 189°C (3mmHg)
Melting Point 91°C to 96°C
Quantity 5 g
UN Number UN3261
Beilstein 1820487
Flash Point >200°C (392°F)
Solubility Information Reacts with water.
Formula Weight 294.86
Percent Purity 96%
Odor Pungent
Sensitivity Moisture sensitive
Chemical Name or Material Chlorotriphenylsilane
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 200-989-0
RTECSNumber VV2720000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only