missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriethylsilane 97.0+%, TCI America™

Catalog No. p-7136350
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05895G 5 g
T058925G 25 g
T0589100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T05895G Supplier TCI America Supplier No. T05895G
Only null left

Chemical Identifiers

CAS 994-30-9
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00000507
InChI Key DCFKHNIGBAHNSS-UHFFFAOYSA-N
Synonym triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
PubChem CID 13819
IUPAC Name chlorotriethylsilane
SMILES CC[Si](Cl)(CC)CC

Specifications

Color Yellow
Boiling Point 149°C
Quantity 5 g
UN Number 2986
Formula Weight 150.72
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorotriethylsilane
EINECSNumber (2)-2041
TSCA Yes