missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotriethylsilane 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 149°C |
| Quantity | 25 g |
| UN Number | 2986 |
| Formula Weight | 150.72 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Chlorotriethylsilane |
Safety and Handling
| EINECSNumber | (2)-2041 |
| TSCA | Yes |