missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriethylsilane, 99%

Catalog No. AC215940250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215940250 25 mL Glass bottle
AC215941000 100 mL Glass bottle
AC215945000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC215940250 Supplier Thermo Scientific Chemicals Supplier No. 215940250

CAS: 994-30-9 | C6H15ClSi | 150.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent

Chemical Identifiers

CAS 994-30-9
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00000507
InChI Key DCFKHNIGBAHNSS-UHFFFAOYSA-N
Synonym triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
PubChem CID 13819
IUPAC Name chloro(triethyl)silane
SMILES CC[Si](Cl)(CC)CC

Specifications

Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 142.0°C to 144.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4300 to 1.4320
Linear Formula (C2H5)3SiCl
Quantity 25 mL
Beilstein 04,627
Fieser 13,165
Merck Index 15,2179
Specific Gravity 0.89
Solubility Information Solubility in water: decomposes
Formula Weight 150.72
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Chlorotriethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 213-615-6

RUO – Research Use Only