missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriethylsilane, 98+%

Catalog No. p-7022664
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1554709 10 g
AAA1554718 50 g
AAA1554730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1554709 Supplier Thermo Scientific Chemicals Supplier No. A1554709

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 994-30-9 | C6H15ClSi | 150.72 g/mol

Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

Solubility
Soluble in aprotic organic solvents.

Notes
Incompatible with bases, strong oxidizing agents, strong acids, alcohols, amines, ammonia, aldehydes and acetone.

Chemical Identifiers

CAS 994-30-9
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00000507
InChI Key DCFKHNIGBAHNSS-UHFFFAOYSA-N
Synonym triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
PubChem CID 13819
IUPAC Name chloro(triethyl)silane
SMILES CC[Si](Cl)(CC)CC

Specifications

Boiling Point 145°C to 147°C
Melting Point ∼-75°C
Quantity 10 g
Flash Point 29°C (84°F)
Odor Pungent
Refractive Index 1.431
Sensitivity Moisture sensitive
Density 0.899
Linear Formula (CH3CH2)3SiCl
UN Number UN2986
Beilstein 1732860
Solubility Information Soluble in aprotic organic solvents.
Formula Weight 150.73
Percent Purity ≥98%
Chemical Name or Material Chlorotriethylsilane
Show More Show Less
Hazard Category H226-H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 213-615-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only