missing translation for 'onlineSavingsMsg'
Learn More

Chlorotri-n-butylsilane, 97%

Catalog No. p-7043920
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0347706 5 g
AAL0347714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0347706 Supplier Thermo Scientific Chemicals Supplier No. L0347706
Only null left

CAS: 995-45-9 | C12H27ClSi | 234.883 g/mol

Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 995-45-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.883
MDL Number MFCD00000509
InChI Key JSQJUDVTRRCSRU-UHFFFAOYSA-N
Synonym silane, tributylchloro, chlorotributylsilane, chlorotri-n-butylsilane, tributyl chloro silane, tri-n-butylchlorosilane, tributylsilyl chloride, tributylchloro-silan, chloro-tri-n-butylsilane
PubChem CID 70448
IUPAC Name tributyl(chloro)silane
SMILES CCCC[Si](CCCC)(CCCC)Cl

Specifications

Boiling Point 93°C to 94°C (4mmHg)
Quantity 5 g
UN Number UN2987
Beilstein 1742537
Flash Point 104°C (219°F)
Solubility Information Reacts with water.
Formula Weight 234.89
Percent Purity 97%
Refractive Index 1.446
Sensitivity Moisture sensitive
Density 0.883
Chemical Name or Material Chlorotri-n-butylsilane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 213-627-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only