Learn More
Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™
$52.25 - $52.25
Chemical Identifiers
| CAS | 133894-48-1 |
|---|---|
| Molecular Formula | C12H24ClF6N3P2 |
| Molecular Weight (g/mol) | 421.733 |
| MDL Number | MFCD00210035 |
| InChI Key | BSCYRXJVGSZNKX-UHFFFAOYSA-N |
| Synonym | pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat |
| PubChem CID | 16211753 |
| IUPAC Name | chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate |
| SMILES | C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5469303
|
Thermo Scientific Chemicals
H5469303 |
1 g |
Each for $52.25
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 133894-48-1 | |
| 421.733 | |
| BSCYRXJVGSZNKX-UHFFFAOYSA-N | |
| 16211753 | |
| C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F |
| C12H24ClF6N3P2 | |
| MFCD00210035 | |
| pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat | |
| chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate |
Specifications
| 133894-48-1 | |
| C12H24ClF6N3P2 | |
| 1 g | |
| 6842341 | |
| pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat | |
| C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F | |
| 421.733 | |
| 421.73 | |
| Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate |
| 145°C | |
| MFCD00210035 | |
| UN1759 | |
| Moisture sensitive | |
| BSCYRXJVGSZNKX-UHFFFAOYSA-N | |
| chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate | |
| 16211753 | |
| ≥98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen