missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorothiazide 97.0+%, TCI America™
$67.11 - $192.00
Chemical Identifiers
| CAS | 58-94-6 |
|---|---|
| Molecular Formula | C7H6ClN3O4S2 |
| Molecular Weight (g/mol) | 295.71 |
| MDL Number | MFCD00058576 |
| InChI Key | JBMKAUGHUNFTOL-UHFFFAOYSA-N |
| Synonym | chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit |
| PubChem CID | 2720 |
| ChEBI | CHEBI:3640 |
| IUPAC Name | 6-chloro-1,1-dioxo-4H-1λâ¶,2,4-benzothiadiazine-7-sulfonamide |
| SMILES | NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl |
Chemical Identifiers
| 58-94-6 | |
| 295.71 | |
| JBMKAUGHUNFTOL-UHFFFAOYSA-N | |
| 2720 | |
| 6-chloro-1,1-dioxo-4H-1λâ¶,2,4-benzothiadiazine-7-sulfonamide |
| C7H6ClN3O4S2 | |
| MFCD00058576 | |
| chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit | |
| CHEBI:3640 | |
| NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl |
Specifications
| 58-94-6 | |
| White-Yellow | |
| MFCD00058576 | |
| chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit | |
| NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl | |
| 295.71 | |
| CHEBI:3640 | |
| ≥97.0% (HPLC,T) | |
| Chlorothiazide |
| 348°C | |
| C7H6ClN3O4S2 | |
| 5 g | |
| JBMKAUGHUNFTOL-UHFFFAOYSA-N | |
| 6-chloro-1,1-dioxo-4H-1λâ¶,2,4-benzothiadiazine-7-sulfonamide | |
| 2720 | |
| 295.71 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DK9450000
TSCA : Yes
Recommended Storage : Refrigerator