missing translation for 'onlineSavingsMsg'
Learn More

Chlorosulfonyl Isocyanate 98.0+%, TCI America™

Catalog No. C088625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C088625G 25 g
C0886100G 100 g
C0886500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C088625G Supplier TCI America Supplier No. C088625G

Chemical Identifiers

CAS 1189-71-5
Molecular Formula CClNO3S
Molecular Weight (g/mol) 141.525
MDL Number MFCD00011608
InChI Key WRJWRGBVPUUDLA-UHFFFAOYSA-N
Synonym chlorosulfonyl isocyanate, chlorosulfonylisocyanate, sulfuryl chloride isocyanate, n-carbonylsulfamyl chloride, chlorosulphonyl isocyanate, sulfurisocyanatidic chloride, n-chlorosulfonyl isocyanate, chlorosulfonyl imino methanone, csi, sulfurisocyanatidoyl chloride
PubChem CID 70918
IUPAC Name N-(oxomethylidene)sulfamoyl chloride
SMILES C(=NS(=O)(=O)Cl)=O

Specifications

Melting Point -80°C
Color Yellow
Boiling Point 107°C
Quantity 25 g
UN Number 3265
Formula Weight 141.53
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorosulfonyl Isocyanate
EINECSNumber (1)-1216
TSCA Yes
Recommended Storage Refrigerator