missing translation for 'onlineSavingsMsg'
Learn More

Chlorosulfonic acid, typically 99%

Catalog No. AA8797730
Change view
Click to view available options
Quantity:
250 g
1 kg
3 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8797730 250 g
AA87977A1 1 kg
AA87977D5 3 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8797730 Supplier Thermo Scientific Chemicals Supplier No. 08797730

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7790-94-5 | ClHO3S | 116.515 g/mol

Chlorosulfonic acid is used as detergent and as an anti-contrail agent. It is used as an intermediate in the production of other substances such as pharmaceuticals and chemicals. It finds application in producing smoke screens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-94-5
Molecular Formula ClHO3S
Molecular Weight (g/mol) 116.515
MDL Number MFCD00011523
InChI Key XTHPWXDJESJLNJ-UHFFFAOYSA-N
Synonym chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909
PubChem CID 24638
IUPAC Name sulfurochloridic acid
SMILES OS(=O)(=O)Cl

Specifications

Appearance Yellow to brown liquid
Color Yellow to Brown
Melting Point -80°C
Boiling Point 158°C
Density 1.753 g/cm3 at 20°C
Odor Odorless
Assay Percent Range typically 99%
Quantity 250 g
UN Number 1754
Physical Form Liquid
Solubility Information Miscible with hydrocarbons.
Formula Weight 116.52
Chemical Name or Material Chlorosulfonic Acid
Show More Show Less
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: I
EINECSNumber 232-234-6
RTECSNumber FX5730000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only