missing translation for 'onlineSavingsMsg'
Learn More

Chlorosulfonic acid, 97%

Catalog No. AC304491000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304491000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC304491000 Supplier Thermo Scientific Chemicals Supplier No. 304491000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7790-94-5 | ClHO3S | 116.515 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-94-5
InChI Key XTHPWXDJESJLNJ-UHFFFAOYSA-N
Synonym chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909
PubChem CID 24638
IUPAC Name sulfurochloridic acid
SMILES OS(=O)(=O)Cl

Specifications

Melting Point − 80.0°C
Boiling Point 151°C to 152°C
Density 1.7500g/mL
Packaging Glass bottle
Assay Percent Range 97%
Quantity 100 mL
Molecular Formula HClO3S
Physical Form Oily Liquid, Fuming In Air
Linear Formula ClSO3H
UN Number 1754
Fieser 01,140; 02,70; 06,121
Merck Index 15, 2168
Solubility Information Solubility in water: reacts with water forming sulfuric and hydrochlori. Other solubilities: de,decomposes in acids,decomposes in alcohol,insoluble in cs2
Molecular Weight (g/mol) 116.52
Formula Weight 116.52
Percent Purity 97%
Chemical Name or Material Chlorosulfonic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 232-234-6

RUO – Research Use Only