missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Chlorosulfonic Acid

Catalog No. 5710241KG
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5710241KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 5710241KG Supplier Sigma Aldrich Supplier No. 5710241KG
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorosulfonic acid is a strong acid that is synthesized on an industrial scale by reacting sulfur trioxide and dry hydrogen chloride gas in an equimolar ratio. It can act as a sulfonating, dehydrating, oxidizing and chlorinating agent. It is reported to be a better electrophilic olefin cyclization agent in comparison to other sulfonic acids. Its infrared spectra in the solid, liquid and gaseous state have been recorded in the region 4000-400cm-1.Chlorosulfonic acid may be used in the following processes: Synthesis of di-n-butyl ammonium chlorosulfonate, a secondary amine ionic liquid.To prepare sulfonated polyethersulfone (SPES) with enhanced hydrophilic property.To attach sulfate group to silk fibroin for improving its anticoagulant ability.As a catalyst to prepare substituted coumarin, via von Pechmann condensation between phenol and +a-ketoester.As a substitute to acid catalyst in Biginelli reaction to prepare 3,4dihydropyrimidines.

Specifications

CAS 7790-94-5
Density 1.753 g/mL at 25°C
Boiling Point 151°C to 152°C (755 mmHg, lit.)
Molecular Formula HClO3S
Refractive Index n20/D 1.433
Linear Formula ClSO3H
MDL Number MFCD00011523
Quantity 1 kg
Molecular Weight (g/mol) 116.52 g/mol
Biological Activity Respiratory System