missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroquine Diphosphate 98.0+%, TCI America™
$132.84 - $1097.34
Chemical Identifiers
| CAS | 50-63-5 |
|---|---|
| Molecular Formula | C18H32ClN3O8P2 |
| Molecular Weight (g/mol) | 515.86 |
| MDL Number | MFCD00069852 |
| InChI Key | QKICWELGRMTQCR-UHFFFAOYNA-N |
| Synonym | chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine |
| PubChem CID | 64927 |
| IUPAC Name | 7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid) |
| SMILES | OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1 |
Chemical Identifiers
| 50-63-5 | |
| 515.86 | |
| QKICWELGRMTQCR-UHFFFAOYNA-N | |
| 64927 | |
| OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1 |
| C18H32ClN3O8P2 | |
| MFCD00069852 | |
| chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine | |
| 7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid) |
Specifications
| 50-63-5 | |
| White-Yellow | |
| MFCD00069852 | |
| chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine | |
| OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1 | |
| 515.86 | |
| 515.86 | |
| Crystalline Powder |
| 199°C | |
| C18H32ClN3O8P2 | |
| 25 g | |
| QKICWELGRMTQCR-UHFFFAOYNA-N | |
| 7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid) | |
| 64927 | |
| ≥98.0% (HPLC,T) | |
| Chloroquine Diphosphate |
Safety and Handling
EINECSNumber : (9)-0279
RTECSNumber : VB2450000
TSCA : No