missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroquine Diphosphate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 199°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 515.86 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Chloroquine Diphosphate |
Safety and Handling
| EINECSNumber | (9)-0279 |
| RTECSNumber | VB2450000 |
| TSCA | No |