missing translation for 'onlineSavingsMsg'
Learn More

Chloropyrazine 98.0+%, TCI America™

Catalog No. p-7124788
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10935G 5 g
C109325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10935G Supplier TCI America Supplier No. C10935G
Only null left

Chemical Identifiers

CAS 14508-49-7
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD00006124
InChI Key GELVZYOEQVJIRR-UHFFFAOYSA-N
Synonym chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
PubChem CID 73277
IUPAC Name 2-chloropyrazine
SMILES ClC1=CN=CC=N1

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 1993
Formula Weight 114.53
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloropyrazine