missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Chloroprocaine hydrochloride
$307.39 - $307.39
Chemical Identifiers
| CAS | 3858-89-7 |
|---|---|
| Molecular Formula | C13H20Cl2N2O2 |
| Molecular Weight (g/mol) | 307.22 |
| InChI Key | SZKQYDBPUCZLRX-UHFFFAOYSA-N |
| IUPAC Name | 2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride |
| SMILES | Cl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465372500
|
Thermo Scientific Chemicals
465372500 |
250 mg |
Each for $307.39
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3858-89-7 | |
| 307.22 | |
| 2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride |
| C13H20Cl2N2O2 | |
| SZKQYDBPUCZLRX-UHFFFAOYSA-N | |
| Cl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl |
Specifications
| 3858-89-7 | |
| C13H20Cl2N2O2 | |
| Solubility aq. soln.: soluble. | |
| Cl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl | |
| 307.22 | |
| Chloroprocaine hydrochloride |
| 173.0°C to 178.0°C | |
| 250 mg | |
| SZKQYDBPUCZLRX-UHFFFAOYSA-N | |
| 2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride | |
| 307.22 |
Safety and Handling
EINECSNumber : 223-371-2
RUO – Research Use Only