missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Chloroprocaine hydrochloride
Supplier: Thermo Scientific™ 465370010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Chloroprocaine hydrochloride | |
| C13H20Cl2N2O2 | |
| Solubility aq. soln.: soluble. | |
| Cl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl | |
| 307.22 |
| 3858-89-7 | |
| 1 g | |
| SZKQYDBPUCZLRX-UHFFFAOYSA-N | |
| 2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride | |
| 307.22 |
Chemical Identifiers
| 3858-89-7 | |
| 307.22 | |
| 2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride |
| C13H20Cl2N2O2 | |
| SZKQYDBPUCZLRX-UHFFFAOYSA-N | |
| Cl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl |
Safety and Handling
EINECSNumber : 223-371-2
RUO â Research Use Only