missing translation for 'onlineSavingsMsg'
Learn More

Chloroplatinic Acid Hexahydrate, ACS Reagent, ≥37.50% Pt basis, Honeywell™

ACS Reagent,≥37.50% Pt basis

$407.45 - $407.45

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045125
View Documents
Honeywell-Fluka
2060831G
1 g Glass Bottle
Each for $407.45
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

18497-13-7
517.89
PIJUVEPNGATXOD-UHFFFAOYSA-J
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
Cl6H14O6Pt
MFCD00149910
platinum(4+) dihydrogen hexahydrate hexachloride

Specifications

18497-13-7
1 g
H2PtCl6 · 6H2O
UN2507
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
517.89
≥37.50% (Pt basis)
Glass Bottle
Powder
Cl6H14O6Pt
MFCD00149910
PIJUVEPNGATXOD-UHFFFAOYSA-J
platinum(4+) dihydrogen hexahydrate hexachloride
517.9g/mol
ACS Reagent
Chloroplatinic Acid Hexahydrate
Videos
Safety and Handling

Safety and Handling

P261-P280-P301 + P310-P305 + P351 + P338-P310

H301-H314-H317-H334

DOTInformation : 8/PGIII

EINECSNumber : 241-010-7

RTECSNumber : TP1510000

SDS
missing translation for 'documents'

missing translation for 'documents'