missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorophenol Red Sodium Salt, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Packaging | Pouch |
| Color | Brown |
| Quantity | 10 g |
| Chemical Name or Material | Chlorophenol Red, Sodium Salt |
Safety and Handling
| ShelfLife | 60 months |