missing translation for 'onlineSavingsMsg'
Learn More

Chlorophenol Red Sodium Salt, Reagent Grade, LabChem™

Catalog No. LC130707
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC130707 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC130707 Supplier LabChem Supplier No. LC130707

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123333-64-2
Molecular Formula C19H11Cl2O5S
Molecular Weight (g/mol) 422.25
MDL Number MFCD00151199
InChI Key QXTPRQZMDKBTAI-UNOMPAQXSA-M
PubChem CID 131846007
IUPAC Name 2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
SMILES OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O

Specifications

CAS Max % 100
Identification Passes Test
Packaging Amber Glass
Linear Formula C19H11Cl2O5SNa
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Color Maroon
Solubility Information Soluble in water
Quantity 10 g
Physical Form Crystals
Formula Weight 445.25
Grade Reagent
Chemical Name or Material Chlorophenol Red Sodium Salt
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Keep container tightly closed.
Store locked up in a well-ventilated place.
Recommended Storage Room Temperature