missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorophenol Red, sodium salt, pure, water soluble

Catalog No. ac216310100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC216310100 10 g Glass bottle
AC216310500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC216310100 Supplier Thermo Scientific Chemicals Supplier No. 216310100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123333-64-2 | C19H11Cl2O5S | 422.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123333-64-2
Molecular Formula C19H11Cl2O5S
Molecular Weight (g/mol) 422.25
MDL Number MFCD00151199
InChI Key QXTPRQZMDKBTAI-UNOMPAQXSA-M
Synonym 3', 3''-Dichlorophenolsulfonephthalein, sodium salt
PubChem CID 131846007
SMILES OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O

Specifications

Melting Point 267.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Extinction (575nm, 0.1N NaOH) ca. 1200
Visual Transition Interval From pH 4.6 (yellow) to pH 7.0 (purple)
Color Brown
Solubility Information Solubility in water: soluble.
Quantity 10 g
Physical Form Crystalline Powder
IUPAC Name 2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol;sodium
Formula Weight 445.25
Grade Pure
Chemical Name or Material Chlorophenol Red, sodium salt
Show More Show Less
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only