missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorophenol Red, pure, indicator grade

Catalog No. AC190040100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190040100 10 g Glass bottle
AC190040500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190040100 Supplier Thermo Scientific Chemicals Supplier No. 190040100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4430-20-0 | C19H12Cl2O5S | 423.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 4430-20-0
Molecular Formula C19H12Cl2O5S
Molecular Weight (g/mol) 423.26
MDL Number MFCD00005877
InChI Key WWAABJGNHFGXSJ-UHFFFAOYSA-N
Synonym CPR, 3', 3''-Dichlorophenylsulfonphthalein
PubChem CID 20486
IUPAC Name 2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1

Specifications

Absorbance 1000 (at lambda max) (calc. on dried substance) (A (1% min./1cm))
Beilstein 19, II, 104
Solubility Information Solubility in water: insoluble
Infrared Spectrum Authentic
Lambda Max 572 to 576nm (in 0.1 N NaOH)
Formula Weight 423.26
Loss on Drying 8% max. (110°C)
Grade Pure
Packaging Glass bottle
Specific Extinction 1000 min. (575nm, in 0.1N NaOH)
Visual Transition Interval From pH 4.8 (yellow) to pH 6.4 (purple)
Color White to Cream
Quantity 10 g
Physical Form Granular Powder
Chemical Name or Material Chlorophenol Red, Pure
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only