missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorophenol Red

CAS: 4430-20-0 | C19H12Cl2O5S | 423.26 g/mol

Supplier:  Thermo Scientific Chemicals B2162314

 View more versions of this product

Catalog No. AAB2162314

Only null left
Explore available promotions

Description

Description

Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.

Solubility
Soluble in water (30 mg/ml), ethanol (80 mg/ml), and methyl cellusolve (50 mg/ml).

Notes
Transition interval: pH 4.8 (yellow) to 6.4 (red)
Specifications

Specifications

Chlorophenol Red
C19H12Cl2O5S
25 g
chlorophenol red, chlorphenol red, chlorphenolsulfonphthalein, dichlorophenolsulfonephthalein, 3,3-bis 3-chloro-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, chlorophenol red indicator, 3',3-dichlorophenolsulfonphthalein, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2-chloro, 3',3'-dichlorophenolsulfonaphthalein, 3',3-dichlorophenol-sulfonphthalein
WWAABJGNHFGXSJ-UHFFFAOYSA-N
2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
20486
4430-20-0
MFCD00005877
354053
Soluble in water (30mg/ml),ethanol (80mg/ml),and methyl cellusolve (50mg/ml).
OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1
423.26
432.28

Chemical Identifiers

4430-20-0
423.26
WWAABJGNHFGXSJ-UHFFFAOYSA-N
20486
OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1
C19H12Cl2O5S
MFCD00005877
chlorophenol red, chlorphenol red, chlorphenolsulfonphthalein, dichlorophenolsulfonephthalein, 3,3-bis 3-chloro-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, chlorophenol red indicator, 3',3-dichlorophenolsulfonphthalein, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2-chloro, 3',3'-dichlorophenolsulfonaphthalein, 3',3-dichlorophenol-sulfonphthalein
2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-619-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only