missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorophenol Red

Catalog No. AAB2162306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2162306 5 g
AAB2162314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2162306 Supplier Thermo Scientific Chemicals Supplier No. B2162306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4430-20-0 | C19H12Cl2O5S | 423.26 g/mol

Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.

Solubility
Soluble in water (30 mg/ml), ethanol (80 mg/ml), and methyl cellusolve (50 mg/ml).

Notes
Transition interval: pH 4.8 (yellow) to 6.4 (red)

Chemical Identifiers

CAS 4430-20-0
Molecular Formula C19H12Cl2O5S
Molecular Weight (g/mol) 423.26
MDL Number MFCD00005877
InChI Key WWAABJGNHFGXSJ-UHFFFAOYSA-N
Synonym chlorophenol red, chlorphenol red, chlorphenolsulfonphthalein, dichlorophenolsulfonephthalein, 3,3-bis 3-chloro-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, chlorophenol red indicator, 3',3-dichlorophenolsulfonphthalein, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2-chloro, 3',3'-dichlorophenolsulfonaphthalein, 3',3-dichlorophenol-sulfonphthalein
PubChem CID 20486
IUPAC Name 2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1

Specifications

Quantity 5 g
Beilstein 354053
Solubility Information Soluble in water (30mg/ml),ethanol (80mg/ml),and methyl cellusolve (50mg/ml).
Formula Weight 432.28
Chemical Name or Material Chlorophenol Red
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-619-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only