missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Polystyrene Resin cross-linked with 2% DVB (200-400mesh) (2.0-2.7mmol/g), TCI America™

Catalog No. C17455G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
C17455G 5 g
1 options

Catalog No. C17455G

Supplier: TCI America C17455G

Only null left

Chemical Identifiers

CAS 55844-94-5
MDL Number MFCD00146406
Synonym Merrifield Resin

Specifications

Quantity 5 g
Chemical Name or Material Chloromethyl Polystyrene Resin cross-linked with 2% DVB (200-400mesh) (2.0-2.7mmol/g)