missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Polystyrene Resin cross-linked with 1% DVB (200-400mesh) (1.6-1.8mmol/g), TCI America™

Catalog No. p-7125100
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C16435G 5 g
C164325G 25 g
2 options

Catalog No. C16435G

Supplier: TCI America C16435G

Only null left

Chemical Identifiers

CAS 55844-94-5
MDL Number MFCD00146406
Synonym Merrifield Resin

Specifications

Quantity 5 g
Chemical Name or Material Chloromethyl Polystyrene Resin cross-linked with 1% DVB (200-400mesh) (1.6-1.8mmol/g)