missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Polystyrene Resin cross-linked with 1% DVB (100-200mesh) (2.0-3.0mmol/g), TCI America™

Catalog No. C17505G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C17505G 5 g
C175025G 25 g
2 options

Catalog No. C17505G

Supplier: TCI America C17505G

Only null left

Chemical Identifiers

CAS 55844-94-5
MDL Number MFCD00146406
Synonym Merrifield Resin

Specifications

Color White-Yellow
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Chloromethyl Polystyrene Resin cross-linked with 1% DVB (100-200mesh) (2.0-3.0mmol/g)