missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloromethyl Ethyl Ether 96.0+%, TCI America™
$165.90 - $1452.10
Chemical Identifiers
| CAS | 3188-13-4 |
|---|---|
| Molecular Formula | C3H7ClO |
| Molecular Weight (g/mol) | 94.54 |
| MDL Number | MFCD00000887 |
| InChI Key | FCYRSDMGOLYDHL-UHFFFAOYSA-N |
| Synonym | chloromethyl ethyl ether, chloromethoxy ethane, ethane, chloromethoxy, chloromethylethylether, ethoxymethyl chloride, ethyl chloromethyl ether, ethoxychloromethane, ether, chloromethyl ethyl, ethoxy methyl chloride |
| PubChem CID | 18523 |
| IUPAC Name | (chloromethoxy)ethane |
| SMILES | CCOCCl |
Chemical Identifiers
| 3188-13-4 | |
| 94.54 | |
| FCYRSDMGOLYDHL-UHFFFAOYSA-N | |
| 18523 | |
| CCOCCl |
| C3H7ClO | |
| MFCD00000887 | |
| chloromethyl ethyl ether, chloromethoxy ethane, ethane, chloromethoxy, chloromethylethylether, ethoxymethyl chloride, ethyl chloromethyl ether, ethoxychloromethane, ether, chloromethyl ethyl, ethoxy methyl chloride | |
| (chloromethoxy)ethane |
Specifications
| 3188-13-4 | |
| 80°C | |
| MFCD00000887 | |
| 2354 | |
| FCYRSDMGOLYDHL-UHFFFAOYSA-N | |
| (chloromethoxy)ethane | |
| 18523 | |
| ≥96.0% (GC) | |
| Chloromethyl Ethyl Ether |
| Yellow | |
| C3H7ClO | |
| 25 g | |
| chloromethyl ethyl ether, chloromethoxy ethane, ethane, chloromethoxy, chloromethylethylether, ethoxymethyl chloride, ethyl chloromethyl ether, ethoxychloromethane, ether, chloromethyl ethyl, ethoxy methyl chloride | |
| CCOCCl | |
| 94.54 | |
| 94.54 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3725
TSCA : Yes