missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)dimethylchlorosilane, 98%

CAS: 1719-57-9 | C3H8Cl2Si | 143.08 g/mol

Supplier:  Thermo Scientific Chemicals A1489914

 View more versions of this product

Catalog No. AAA1489914


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

110°C to 113°C
1719-57-9
C3H8Cl2Si
MFCD00000875
1731933
chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane
C[Si](C)(Cl)CCl
143.08
143.09
Acrid
Moisture sensitive
(Chloromethyl)dimethylchlorosilane
25 g
ClCH2Si(CH3)2Cl
UN2985
21°C (69°F)
ITKVLPYNJQOCPW-UHFFFAOYSA-N
chloro-(chloromethyl)-dimethylsilane
74393
98%
1.437
1.082

Chemical Identifiers

1719-57-9
143.08
ITKVLPYNJQOCPW-UHFFFAOYSA-N
74393
C[Si](C)(Cl)CCl
C3H8Cl2Si
MFCD00000875
chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane
chloro-(chloromethyl)-dimethylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 217-006-6

RTECSNumber : VV2095000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only