missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)cyclopropane 95.0+%, TCI America™

Catalog No. C24695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24695G 5 g
C246925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24695G Supplier TCI America Supplier No. C24695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5911-08-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00001307
InChI Key ZVTQWXCKQTUVPY-UHFFFAOYSA-N
Synonym chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane
PubChem CID 80013
IUPAC Name (chloromethyl)cyclopropane
SMILES ClCC1CC1

Specifications

Melting Point -91°C
Color Yellow
Boiling Point 88°C
Quantity 5 g
UN Number 1993
Formula Weight 90.55
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (Chloromethyl)cyclopropane