missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)cyclopropane, 97%

Catalog No. AAL0698606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0698606 5 g
AAL0698614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0698606 Supplier Thermo Scientific Chemicals Supplier No. L0698606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5911-08-0 | C4H7Cl | 90.55 g/mol

(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4?-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4′-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and heat.

Chemical Identifiers

CAS 5911-08-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00001307
InChI Key ZVTQWXCKQTUVPY-UHFFFAOYSA-N
Synonym chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane
PubChem CID 80013
SMILES ClCC1CC1

Specifications

Melting Point -91°C
Density 0.98
Boiling Point 87°C to 89°C
Flash Point −1°C (30°F)
Odor Recognizable
Refractive Index 1.436
Quantity 5 g
UN Number UN1993
Beilstein 635697
Solubility Information Not miscible in water.
IUPAC Name chloromethylcyclopropane
Formula Weight 90.55
Percent Purity 97%
Chemical Name or Material (Chloromethyl)cyclopropane
Show More Show Less
Hazard Category H225-H302
Hazard Statement GHS H Statement
H225-H302-H315-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 227-632-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only