missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl chlorosulfate, 97%

Catalog No. p-3735987
Change view
Click to view available options
Quantity:
5 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330800050 5 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC330800050 Supplier Thermo Scientific Chemicals Supplier No. 330800050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 49715-04-0 | CH2Cl2O3S | 164.984 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49715-04-0
MDL Number MFCD00041547
InChI Key PJBIHXWYDMFGCV-UHFFFAOYSA-N
Synonym chloromethyl chlorosulfate, chloromethyl chlorosulphate, chloromethylchlorosulfate, chloromethyl chlorosulfonate, chloromethyl chloranesulfonate, unii-n615783bnf, chloromethyl sulfochloridate, chlorosulfuric acid, chloromethyl ester, chloro chlorosulfonyloxy methane, chlorosulfuric acid chloromethyl ester
PubChem CID 2733556
IUPAC Name chloro(chlorosulfonyloxy)methane
SMILES C(OS(=O)(=O)Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.6000g/mL
Boiling Point 45.0°C to 50.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Molecular Formula CH2Cl2O3S
Refractive Index 1.4460 to 1.4480
Quantity 5 g
Beilstein 01, III, 2593
Specific Gravity 1.6
Merck Index 15, 2147
Solubility Information Solubility in water: reacts.
Molecular Weight (g/mol) 164.99
Formula Weight 164.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chloromethyl chlorosulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 256-442-1

RUO – Research Use Only