missing translation for 'onlineSavingsMsg'
Learn More

Chloroiodomethane (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. C117925G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C117925G 25 g
C1179100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C117925G Supplier TCI America Supplier No. C117925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 593-71-5
Molecular Formula CH2ClI
Molecular Weight (g/mol) 176.381
MDL Number MFCD00001078
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane
SMILES C(Cl)I

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 g
Formula Weight 176.38
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloroiodomethane (stabilized with Copper chip)