Learn More
Chloroiodomethane, 98%, stabilized
CAS: 593-71-5 | CH2ClI | 176.38 g/mol
Supplier: Thermo Scientific Chemicals 222151000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
| Chloroiodomethane | |
| Pink-Red to Yellow | |
| 108.0°C to 109.0°C | |
| 97.5% min. (GC) | |
| CH2ClI | |
| ClCH2I | |
| 01,71 | |
| 2.42 | |
| Solubility in water: insoluble. Other solubilities: soluble in many organic solvents | |
| C(Cl)I | |
| 176.38 | |
| 11644 | |
| 98% |
| 593-71-5,7440-22-4 | |
| 2.4200g/mL | |
| Authentic | |
| Glass bottle | |
| 1.5812 to 1.5832 | |
| 100 mL | |
| 10,89; 17,115 | |
| chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@ | |
| PJGJQVRXEUVAFT-UHFFFAOYSA-N | |
| chloro(iodo)methane | |
| Copper | |
| 176.38 | |
| Liquid |
Chemical Identifiers
| 593-71-5 | |
| 176.38 | |
| chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@ | |
| chloro(iodo)methane |
| CH2ClI | |
| PJGJQVRXEUVAFT-UHFFFAOYSA-N | |
| 11644 | |
| C(Cl)I |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 209-804-8
RUO – Research Use Only