missing translation for 'onlineSavingsMsg'
Learn More

Chloroiodomethane, 98%, stabilized

Catalog No. AC222150250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222150250 25 mL Glass bottle
AC222151000 100 mL Glass bottle
AC222155000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC222150250 Supplier Thermo Scientific Chemicals Supplier No. 222150250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 593-71-5 | CH2ClI | 176.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 593-71-5 , 7440-22-4
Molecular Formula CH2ClI
Molecular Weight (g/mol) 176.38
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane
SMILES C(Cl)I

Specifications

Color Pink-Red to Yellow
Density 2.4200g/mL
Boiling Point 108.0°C to 109.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5812 to 1.5832
Linear Formula ClCH2I
Quantity 25 mL
Beilstein 01,71
Fieser 10,89; 17,115
Specific Gravity 2.42
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
Stabilizer Copper
Formula Weight 176.38
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloroiodomethane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 209-804-8

RUO – Research Use Only